missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Stand 4 in. Riser (short)
Click to view available options
Height (English):
4 in.
Unit Size:
Each
Description
Positioning bases, increases the height of the shaker for easier access, for Innova™ 44/44R, 10.2 cm, short
Specifications
Specifications
| Product Type | Short Stand |
| Height (English) | 4 in. |
| Height (Metric) | 10.2 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction