Learn More
Integra Biosciences™ Stand
Product Code. 10328811
Change view
Click to view available options
:
For use with: Parking vacoboy hand operator
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10328811 For use with: Parking vacoboy hand operator
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10328811 Supplier Integra Biosciences™ Supplier No. 155501
VACUBOY hand operator set comprises of: VACUBOY hand operator, stand, rubber adapter for Pasteur pipettes, 1 channel stainless steel aspiration adapter 40mm, 8 channel stainless steel aspiration adapter 40mm, 1 channel plastic aspiration adapter for disposable tips, 1 channel plastic aspiration adapter for disposable tips with ejector, 8 channel plastic aspiration adapter for disposable tips with ejector. Can be used with VACUSAFE comfort vacuum system. Full range of accessories available. Enables liquid residues to be disposed of reliably, precisely and conveniently:
  • With a precisely dosed volume
  • Without loss of drops
  • Without risk of contamination
  • Without causing tiredness
  • Energy saving, the manual operator - the heart of the system - can be operated as a single or 8 channel aspirator
  • All parts in contact with fluids are autoclavable
Specifications
Description Vacuboy bench stand