Learn More
Schneider Gerd™ Stainless Steel Tweezer
Click to view available options
Length (Metric):
105 mm
115 mm
Tip Style:
Grooved
Spade
Unit Size:
Each
Description
- Stainless steel tweezers
- Available with straight or curved tips
- Ideal for dissection and general lab applications
Specifications
Specifications
| Tip Style | Grooved |
| Material | Stainless Steel |
| Length (Metric) | 105 mm |
| Product Type | Tweezers |
| For Use With (Application) | For Laboratory use |