Learn More
Gilac™ Stainless Steel Trolley for Large Volume Container
Description
- Trolley equiped with 125 mm diameter wheels in white PA (2 fixed and 2 pivoting with brakes).
- Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
- Makes it easier to work at breast height.
Specifications
Specifications
| Type | Trolley |
| Holds | Large Volume Containers |
| Load Bearing Capacity (Metric) | 200 kg |
| Material | Stainless Steel |
| Caster Diameter (Metric) | 125 mm |
| For Use With (Equipment) | 220 L Container |
| Height (English) | 28.3 in. |
| Height (Metric) | 720 mm |