Learn More
RETSCH Stainless-Steel Test Sieves, 450 mm Dia., in Micrometer Pore Sizes with ASTM E 11
Product Code. p-8000557 Shop All Retsch Products
Click to view available options
Pore Size:
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
106 μm
125 μm
150 μm
180 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
Unit Size:
Each
Product Code. 11934702
Brand: RETSCH 60.169.000300
Only null left
Specifications
Quantity 1 Each
Pore Size 300 μm
Type Test Sieve
Certifications/Compliance ASTM E 11
For Use With (Equipment) Vibratory Shaker AS 450
Includes Certificate indicating Compliance
Height (Metric) 100mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 450 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel