Learn More
asecos™ Stainless Steel Small Parts Cleaner
Product Code. p-200003688 Shop All asecos Products
Change view
Click to view available options
Capacity (Metric):
2 L
4 L
8 L
Color:
Gray
Yellow
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color unitSize
17876386 2 L Yellow Each
17826466 4 L Yellow Each
17886346 2 L Gray Each
17836556 4 L Gray Each
17856556 8 L Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17876386 Supplier asecos™ Supplier No. 40445
The Stainless Steel Small Parts Cleaner is ideal for cleaning or degreasing small parts.
  • With recessed grip
  • Spring-loaded immersion sieve incl. flame arrester, grounding connection
Specifications
Product Type Parts Cleaner
Capacity (English) 0.5 gal.
Capacity (Metric) 2 L
Color Yellow
Material Steel
Width (English) 10.39 in.
Width (Metric) 264.00 mm
Height (English) 3.74 in.
Height (Metric) 95.00 mm
Weight (English) 5.64 lb.
Weight (Metric) 2.56 kg