Learn More
VACUUBRAND™ Stainless Steel Small KF Flange With Male Ground Joint
Click to view available options
Diameter (Metric) Inside:
10 mm
25 mm
40 mm
Unit Size:
Each
Specifications
Specifications
| Diameter (Metric) Inside | 10 mm |
| Material | Stainless Steel |
| Quantity | 1 Each |
| Size | Small |
| Size (Metric) | 33 x 23 x 13.2 mm |
| Type | Small KF Flange |