Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for UF160, 15 mm
Product Code. 17539025 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF160 Oven
Unit Size:
Each
Product Code. 17539025
Brand: Memmert™ UF160B02073
Only null left
Add to basket
Add to basket
Non-perforated slide-in drip trays constructed of stainless steel.
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specifications
For Use With (Equipment) UF160 Oven
Type Drip Tray
Material Stainless Steel
Height (Metric) Rim: 15 mm