Learn More
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh
Made from stainless steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has an 100mm diameter.
28.50 EUR - 213.00 EUR
Specifications
Diameter (Metric) Outer | 100 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Woven Wire |
Quantity | 1 Each |
Type | Sieve |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
11572143
|
Fisherbrand™
|
106μm |
45.00 EUR
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11582163
|
Fisherbrand™
|
1.25mm |
43.95 EUR
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
11592163
|
Fisherbrand™
|
1.4mm |
45.25 EUR
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
11592143
|
Fisherbrand™
|
150μm |
44.60 EUR
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
11502153
|
Fisherbrand™
|
160μm |
43.95 EUR
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
11512153
|
Fisherbrand™
|
180μm |
44.60 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
11522173
|
Fisherbrand™
|
2mm |
47.90 EUR
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
11522153
|
Fisherbrand™
|
200μm |
28.50 EUR
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
11532153
|
Fisherbrand™
|
212μm |
43.95 EUR
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11542173
|
Fisherbrand™
|
2.5mm |
45.25 EUR
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
11582133
|
Fisherbrand™
|
25μm |
213.00 EUR
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11552173
|
Fisherbrand™
|
2.8mm |
52.05 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11552153
|
Fisherbrand™
|
300μm |
43.95 EUR
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11562173
|
Fisherbrand™
|
3.35mm |
51.90 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11592133
|
Fisherbrand™
|
38μm |
59.90 EUR
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
11582153
|
Fisherbrand™
|
400μm |
44.45 EUR
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
11572153
|
Fisherbrand™
|
355μm |
43.95 EUR
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
|||||
11592153
|
Fisherbrand™
|
425μm |
30.15 EUR
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
11502163
|
Fisherbrand™
|
500μm |
45.25 EUR
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
11512143
|
Fisherbrand™
|
53μm |
51.90 EUR
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11532143
|
Fisherbrand™
|
75μm |
45.25 EUR
Each |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
11532163
|
Fisherbrand™
|
710μm |
43.95 EUR
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||||
11512163
|
Fisherbrand™
|
600μm |
43.95 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11522163
|
Fisherbrand™
|
630μm |
43.95 EUR
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
11542163
|
Fisherbrand™
|
800μm |
43.95 EUR
Each |
Estimated Shipment: 02-07-2024
Log in to see stock. |
|||||
11542143
|
Fisherbrand™
|
80μm |
43.95 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11552143
|
Fisherbrand™
|
90μm |
43.95 EUR
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11552163
|
Fisherbrand™
|
850μm |
49.25 EUR
Each |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
11562153
|
Fisherbrand™
|
0.315mm |
43.95 EUR
Each |
Estimated Shipment: 15-07-2024
Log in to see stock. |
|||||
11572163
|
Fisherbrand™
|
1.18mm |
47.90 EUR
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11502173
|
Fisherbrand™
|
1.6mm |
45.25 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11512173
|
Fisherbrand™
|
1.7mm |
46.60 EUR
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
11532173
|
Fisherbrand™
|
2.36mm |
46.60 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
11562143
|
Fisherbrand™
|
100μm |
43.95 EUR
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
11582143
|
Fisherbrand™
|
125μm |
44.70 EUR
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
11562163
|
Fisherbrand™
|
1mm |
47.25 EUR
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
11502143
|
Fisherbrand™
|
45μm |
58.55 EUR
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
11522143
|
Fisherbrand™
|
63μm |
46.60 EUR
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
|||||
11542153
|
Fisherbrand™
|
250μm |
44.20 EUR
Each |
Estimated Shipment: 26-04-2024
Log in to see stock. |
|||||
Description
- 100mm diameter test sieves
- Stainless steel frame
- Woven wire mesh
- Supplied with Certificate of Conformity and record card
Specifications
100 mm | |
Woven Wire | |
Sieve |
Stainless Steel | |
1 Each | |
Certificate of conformity and record card |