Learn More
Hanna Instruments™ Stainless Steel Probe
Click to view available options
For Use With (Application):
Measuring & penetration of semisolid
Measuring liquids & air
Unit Size:
Each
Specifications
Specifications
| Description | Probe |
| Material | Stainless Steel |
| Sensor Types | PT 100 |
| For Use With (Application) | Measuring liquids & air |
| Accuracy | ±0.25°C |
| Temperature Range (Metric) | -30°C to +350°C |
| Length (Metric) | 1 m |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur