Learn More
Hanna Instruments™ Stainless Steel Probe
Product Code. 10316061
Click to view available options
For Use With (Application):
Measuring & penetration of semisolid
Measuring liquids & air
Unit Size:
Each
Product Code. 10316061
Brand: Hanna Instruments™ HI768P
Specifications
Description Probe
Material Stainless Steel
Sensor Types PT 100
For Use With (Application) Measuring & penetration of semisolid
Accuracy ±0.25°C
Response Time 30 sec.
Temperature Range (Metric) -30°C to +350°C
Length (Metric) 1 m