Learn More
Bochem™ Stainless Steel Paper Scissors
Click to view available options
Length (Metric):
175 mm
200 mm
250 mm
275 mm
300 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Paper Scissors |
| Length (Metric) | 300 mm |
| Material | Stainless Steel |
| Tip Style | Sharp/Sharp |