Learn More
Cole-Parmer™ Stainless Steel Panel-Mount Solar Powered Thermometers
Shop All Cole Parmer Products
Click to view available options
Temperature Range (Metric):
-40°C to +70°C
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Perfect For Use In Freezers and Walk-In Coolers |
| Material | Stainless Steel |
| Product Type | Panel-Mount Solar Powered Thermometer |
| Temperature Range (Metric) | -40°C to +70°C |