Learn More
Sonics Materials™ Stainless Steel Optional Temperature Probe
Product Code. 11723129
Click to view available options
For Use With (Equipment):
VCX
Unit Size:
Each
Product Code. 11723129
Brand: Sonics Materials™ 83000060
Only null left
Specifications
Product Type Optional Temperature Probe
Material Stainless Steel
For Use With (Application) Allows sample temperature to be monitored up to 100°C
For Use With (Equipment) VCX
Packaging Quantity 1 Ea.