Learn More
Sartorius™ Stainless Steel OIML Class F2 Calibration Weight Without DAkkS Calibration Certificate
Product Code. p-8006442 Shop All Sartorius Products
Click to view available options
Mass:
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
Each
Product Code. 10591541
Brand: Sartorius™ YCW514800
Highly polished steel calibration weights from 100g to 10kg
To OIML Class E2, F1 and F2 specification which includes DKD (Deutscher Kalibrierdienst accredited by the PTB) certificate (UKAS Equivalent).
TRUSTED_SUSTAINABILITY
Specifications
Case Material Plastic
Mass 100 g