Learn More
Sampling Systems™ Stainless Steel Mini-Drums without Toggle Clamps
Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
5 L
6 L
8 L
10 L
12.5 L
15 L
Unit Size:
Each
Description
- Manufactured from durable 304 grade stainless steel
- Crevice-free interior
- Neat recessed fitting lid
- Complete with a swing carrying handle
- Flat bottom with chime ring to give added protection
- Available in a wide range of capacities to fit your specific needs
Specifications
Specifications
| Closure Included | Yes |
| Product Type | Mini Drum |
| Closure Type | Recessed Lid |
| Capacity (Metric) | 6 L |
| For Use With (Application) | Multi-purpose Storage |
| Material | Stainless Steel |
| Includes | Swing Handle |
| Height (Metric) | 280 mm |
| Diameter (Metric) Outer | 175 mm |