Learn More
Sampling Systems™ Stainless Steel Mini-Drums without Toggle Clamps
Product Code. 15819671 Shop All Sampling Systems Products
Click to view available options
Capacity (Metric):
1 L
2 L
3 L
5 L
6 L
8 L
10 L
12.5 L
15 L
Unit Size:
Each
Product Code. 15819671
Brand: Sampling Systems™ A43412
These Mini-Drums without Toggle Clamps are excellent multi-purpose containers for a wide variety of applications in the laboratory.
  • Manufactured from durable 304 grade stainless steel
  • Crevice-free interior
  • Neat recessed fitting lid
  • Complete with a swing carrying handle
  • Flat bottom with chime ring to give added protection
  • Available in a wide range of capacities to fit your specific needs
Specifications
Closure Included Yes
Product Type Mini Drum
Closure Type Recessed Lid
Capacity (Metric) 12.5 L
For Use With (Application) Multi-purpose Storage
Material Stainless Steel
Includes Swing Handle
Height (Metric) 285 mm
Diameter (Metric) Outer 237 mm