Learn More
Schneider Gerd™ Stainless Steel Micro-Spoon Spatula
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Description
- Micro-spoons collect and transport dry substances or liquids
- Spatulas are ideal for working with powders, stirring and mixing
- Perfect for working with miniscule amounts of chemicals
- Two different end shapes, offer increased versatility
- Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
| Quantity | 1 Ea. |
| Shape | Spoon |
| Product Type | Micro Spoon Spatula |
| Width (Metric) | 6 mm |
| Material | Stainless Steel |
| Length (Metric) | 150 mm |