Learn More
Elma™ Stainless Steel Holder Set
Description
- Rilsan-coated holder hooks
- Holder hooks with 10 hooks
- Holder hooks are ideal for holding jewelry and watch parts
- Holder shaft with 4 shafts
- Holder shaft is ideal for holding impression tray
Specifications
Specifications
| Type | Modular Basket System |
| For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
| Includes | Holder Shaft, Holder Hook, 5 Silicone Holders |
| Material | Stainless Steel |