missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Stainless Steel, XM-32, M152, S64152, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Stainless Steel, XM-32, M152, S64152, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Boron/Titanium/Tin/Oxygen/Phosphorus/Cobalt/Niobium/Nitrogen/Copper/Carbon/Silicon/Vanadium/Manganese/Molybdenum/Nickel/Chromium/Tantalum/Aluminium/Tungsten/Zirconium |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.001 %/Titanium 0.0011 %/Tin 0.004 %/Oxygen 0.014 %/Phosphorus 0.016 %/Cobalt 0.021 %/Niobium 0.022 %/Nitrogen 0.035 %/Copper 0.06 %/Carbon 0.11 %/Silicon 0.23 %/Vanadium 0.29 %/Manganese 0.71 %/Molybdenum 1.61 %/Nickel 2.62 %/Chromium 11.3 %/Tantalum (0.001) %/Aluminium (0.004) %/Tungsten (0.01) %/Zirconium <0.005 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction