Learn More
Hamilton™ Stainless Steel HC-75 (Pb2+) Cation Exchange Column
Specifications
Specifications
| Diameter (Metric) | 7.8 mm |
| Length (Metric) | 305 mm |
| Max. Pressure | 400 psi |
| Particle Size | 9 μm |
| Pore Size | Gel-Type |
| Quantity | 1 Ea. |
| USP Type | L34 |
| Stationary Phase | HC-75 (Pb2+) |
| Packing Material | PSDVB/Sulfonic Acid |
| Material | Stainless Steel |
| Show More |