Learn More
Bochem™ Stainless Steel Funnel
Product Code. 10018510
Click to view available options
Diameter (Metric)Top:
100mm
150mm
200mm
250mm
Height (Metric):
125 mm
155 mm
180 mm
190 mm
220 mm
260 mm
Unit Size:
Each
Product Code. 10018510
Brand: Bochem™ 8843
Used for a wide range of applications. Bochem™ Stainless Steel Funnel has a conical shape.
  • Handle 
  • With air canal
  • Tube conical shape
  • 18/10 Stainless steel
Specifications
Diameter (Metric)Top 150mm
Material Stainless Steel
Height (Metric) 190 mm