Learn More
Usbeck Laborgeraete™ Stainless Steel Forceps
Product Code. 10005611
Click to view available options
Length (Metric):
105 mm
115 mm
130 mm
145 mm
160 mm
200 mm
Design:
Curved
Straight
Unit Size:
Each
Product Code. 10005611
Brand: Usbeck Laborgeraete™ 3013
Specifications
Tip Style Blunt
No. per Pack 1 Pack
Design Straight
Material Stainless Steel
Length (Metric) 145 mm
Dimensions (L x W) 145 L mm
Type Forceps