Learn More
Schneider Gerd™ Stainless Steel Double Spoon Micro-spatula
Click to view available options
Length (Metric):
150 mm
210 mm
Unit Size:
Each
Description
- Stainless Steel Double Spoon Micro-spatula
- Spatulas are ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
- Two end shapes offer increased versatility
- Polished stainless steel implements are effortlessly cleaned and corrosion resistant
Specifications
Specifications
| Quantity | 1 Ea. |
| Shape | Double Spoon |
| Product Type | Double Spoon Micro Spatula |
| Material | Stainless Steel |
| Length (Metric) | 210 mm |