Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class F2
Shop All Fisherbrand Products
Click to view available options
Weight (Metric):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
Unit Size:
1 unit
Description
- Class F2 stainless steel calibration weight
- Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
- In compliance with OIML R111
- Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
| Classification | F2 |
| Weight (Metric) | 1 g to 5 kg |
| Mass | 1 g to 5 kg |
| Shape | Cylindrical |