Learn More
RETSCH Stainless Steel Comfort Grinding Jars
Shop All Retsch Products
Click to view available options
Capacity (Metric):
12 mL
25 mL
80 mL
Unit Size:
Each
Specifications
Specifications
| Product Type | Comfort Grinding Jar |
| Capacity (Metric) | 25 mL |
| For Use With (Equipment) | For PM 100, PM 400 |
| Material | Stainless Steel |