Description
Clamp (hose) 1/2 in. (0.43-0.81 0.0)
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Material | Stainless Steel |
| Sterility | Non-sterile |
| No. per Pack | 4Pack |
| Quantity | 4Pack |
| Type | Laboratory Filter Holders or Cyclones |
| Diameter (English) | 0.5 in. |
| Diameter (Metric) Inside | 1.25 cm |