Learn More
Socorex™ Stainless Steel Bent Drenchers
36.45 EUR - 46.05 EUR
Specifications
Product Type | Bent Drencher |
---|---|
Autoclavable | Autoclavable |
Material | Stainless Steel |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11843203
|
Socorex™
376.1251B |
51 mm |
46.05 EUR
Pack of 3 |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11883193
|
Socorex™
376.0925B |
25 mm |
36.45 EUR
Pack of 3 |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Bent Drencher | |
Autoclavable | |
Stainless Steel |