Learn More
VACUUBRAND™ Stainless Steel Ball Valve
Click to view available options
Type:
Ball Valve VKE 16
Ball Valve VKE 25
Ball Valve VKE 40
Size:
Diameter: 16 mm
Diameter: 25 mm
Diameter: 40 mm
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel |
| Quantity | 1 Each |
| Size | Diameter: 16 mm |
| Type | Ball Valve VKE 16 |