missing translation for 'onlineSavingsMsg'
Learn More

Stainless Steel, AISI 17-4 PH, S17400, 50g, ARMI∣MBH

Product Code. 30002549
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002549 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30002549 Supplier ARMI/MBH Supplier No. MBH13X PH17400 AC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.

Stainless Steel, AISI 17-4 PH, S17400, 50g
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tungsten/Carbon/Phosphorus/Nitrogen/Molybdenum/Vanadium/Niobium/Silicon/Manganese/Copper/Nickel/Chromium
CAS 7439-89-6
Quantity 50 g
Form Pieces
Concentration or Composition (by Analyte or Components) Tungsten 0.0056 %/Carbon 0.02 %/Phosphorus 0.02 %/Nitrogen 0.0342 %/Molybdenum 0.061 %/Vanadium 0.112 %/Niobium 0.184 %/Silicon 0.349 %/Manganese 0.829 %/Copper 3.09 %/Nickel 4.52 %/Chromium 15.74 %
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.