missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Hypodermic Needles

Designed for patient comfort in a broad array of sizes. B Braun™ Hypodermic Needles provide a smooth surface with light silicone coating designed to minimize discomfort.

Brand:  B Braun™ 4665503

Additional Details : Weight : 0.13050kg

 View more versions of this product

Product Code. 10156590

  • 7.90 EUR / Pack of 100
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

 Unique, highly automated grinding and polishing process and low-friction lubrication is designed to achieve unequaled sharpness and ease of penetration. Transparent hub design provides high visibility to detect flashback.
  • Color-coded hubs conform to ISO 6009 for easy identity
  • Wide range of diameters and lengths
  • Gauges from 18G to 27G
  • Ultra-sharp, tri-beveled stainless steel designed to result in minimal pain

Specifications

Specifications

Polypropylene (Hub), Stainless Chromium Nickel Steel
21 G
For minimal pain upon puncture, single use
Needle
Ultra-sharp Long Tri-bevel
Green
0.8 mm
Yes
50 mm
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers