Learn More
B Braun™ Sterican™ Single-use Tri-beveled Needles
Product Code. 11353280
Click to view available options
Needle Gauge:
23 G
Unit Size:
Pack of 100
Product Code. 11353280
Brand: B Braun™ 4657667
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from stainless chromium-nickel steel with extremely smooth surface and light silicone coating. B Braun™ Sterican™ Single-use Tri-beveled Needles provide minimal pain upon puncture. Latex-free/PVC-free
  • Transparent Luer Lock plastic hub made of polypropylene
  • Colour-coded hub identifies the outer diameter of the needle, conforms to ISO 6009
  • Needle made of stainless chromium nickel steel with extremely smooth surface and light silicone coating
  • 3-facet bevel for minimal pain upon puncture; bevel length according to indication
  • Latex-free/PVC-free
Specifications
Certifications/Compliance ISO 7864 and DIN 13097
Material Stainless Chromium Nickel Steel
Needle Gauge 23 G
Diameter (Metric) Outer 0.6 mm
For Use With (Application) Intravenous, intramuscular injections
Latex Free Yes
Product Type Disposal Needle
Disposable Single-use
Quantity 100/Pk.
Connection Type Luer Lock