missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAG2 Monoclonal Antibody (KT76), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen™ MA555997
This item is not returnable.
View return policy
Description
STAG2 is a component of the cohesin complex. This complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The cohesin complex may also play a role in spindle pole assembly during mitosis.
Specifications
| STAG2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| O35638, Q8N3U4 | |
| STAG2 | |
| Recombinant mouse Stag2 (proprietary range information). | |
| 200 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgM |
| Western Blot | |
| KT76 | |
| Unconjugated | |
| STAG2 | |
| 9230105L23Rik; B230112I07Rik; bA517O1.1; cohesin subunit SA-2; DKFZp686P168; DKFZp781H1753; FLJ25871; nuclear protein SA2; OTTHUMP00000024338; OTTHUMP00000043514; OTTHUMP00000043515; OTTHUMP00000081754; OTTHUMP00000081755; RGD1562042; RP11-517O1.1; SA2; SA-2; Sap2; SCC3 homolog 2; SCC3B; STAG2; Stromal antigen 2 | |
| Rat | |
| Protein L | |
| RUO | |
| 10735, 20843 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction