Learn More
Invitrogen™ ST3GAL5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530289
Description
Recommended positive controls: A431, Jurkat, Raji, mouse brain, U87-MG. Predicted reactivity: Mouse (91%), Rat (90%), Sheep (95%), Rhesus Monkey (98%), Chimpanzee (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintenance of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene.
Specifications
| ST3GAL5 | |
| Polyclonal | |
| Unconjugated | |
| ST3GAL5 | |
| [a]2; 3S-T; alpha 2,3-sialyltransferase V; alpha2,3-sialyltransferase; CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; Ganglioside GM3 synthase; GM3 synthase; GM3S; GM3-specific sialytransferase; lactosylceramide alpha-2,3-sialyltransferase; lactosylceramide alpha-2,3-sialyltransferas-like protein; mST3Gal V; SATI; Sialyltransferase 9; sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase); sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; GM3 synthase); Siat9; SIATGM3S; ST3 beta-galactoside alpha-2,3-sialyltransferase 5; ST3 beta-galactoside alpha-23-sialyltransferase 5; ST3Gal V; ST3GAL5; ST3GalV; ST3GAL-V; UNQ2510/PRO5998 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20454, 8869 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol; pH 7 | |
| O88829, Q9UNP4 | |
| ST3GAL5 | |
| Recombinant protein encompassing a sequence within the center region of human ST3GAL5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.