missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ST2 (IL-33R) ELISA Kit
Description
The Human ST2 (IL-33R) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of human ST2 (IL-33R).
IL-33R, also known as ST2, is a receptor for IL-33. It is a member of the IL-1 receptor family of innate receptors. Its extracellular domain consists of 3 Ig-like domains that directly interact with its co-receptor IL-1RAcP, and the ligand alarmin, IL-33. IL-33 ligation results in the association of IL-33R complex with its adaptor proteins MyD88, IRAK1, IRAK4 and TRAF6, and activation of the NF-kB, and MAPK pathways. This, in turn, instigates the release of chemokines and cytokines such as IL-5, IL-6, IL-8, and IL-13. Two most common isoforms of IL-33R that result from alternative splicing are ST2L - the membrane bound form, and ST2S - the soluble form. The membrane bound form can be released from the cell surface by proteolytic cleavage. The soluble forms have been suggested to act as decoys dampening the IL-33 signaling. IL-33R is primarily expressed by mast cells, basophils, and eosinophils. In mouse, IL-33R expression has been shown on ILC2 and Th2 cells. IL-33R signaling has been implicated in the number of immune conditions, including allergies, arthritis, atherosclerosis and sepsis.
Specifications
Specifications
| Accession Number | DER4; FIT1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; Il1rl1; IL-1 RL1; IL33R; IL-33 R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; Lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33 R; soluble IL 33 Receptor; soluble IL 33 R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2 |
| Assay Range | 4.7 to 300 pg/mL |
| Assay Sensitivity | 4.6 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Cell Lysates, Plasma, Serum |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 9173 |
| Gene Symbol | IL1RL1 |
| Interassay CV | 3.5% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?