missing translation for 'onlineSavingsMsg'
Learn More

ST-836 hydrochloride, MedChemExpress

Product Code. p-200096047
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456388 10 mg 10mg
16452497 50 mg 50mg
16448977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16456388 Supplier Medchem Express Supplier No. HY15238A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ST-836 hydrochloride (compound 34) is a potent dopamine receptor ligand with Ki values of 4.5 nM, 132 nM for D3 and D2, respectively. ST-836 hydrochloride has the potential for Parkinson’s disease.

ST-836 hydrochloride (compound 34) is a potent dopamine receptor ligand with Ki values of 4.5 nM, 132 nM for D3 and D2, respectively. ST-836 hydrochloride has the potential for Parkinson’s disease.

Specifications

Chemical Name or Material ST-836 hydrochloride
CAS 1415564-68-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H35ClN4OS
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (221.70 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (110.85 mM; Need ultrasonic)
SMILES [H]Cl.CCCN(CCN1CCN(C2=CC=CC=C2OC)CC1)C3CCC4=C(SC=N4)C3
Molecular Weight (g/mol) 451.07
Formula Weight 451.07
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.