Learn More
ST-836 hydrochloride, MedChemExpress
Product Code. 16456388
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16456388
Brand: Medchem Express HY15238A
MedChemExpress ST-836 hydrochloride (compound 34) is a potent dopamine receptor ligand with Ki values of 4.5 nM, 132 nM for D3 and D2, respectively. ST-836 hydrochloride has the potential for Parkinson’s disease.
ST-836 hydrochloride (compound 34) is a potent dopamine receptor ligand with Ki values of 4.5 nM, 132 nM for D3 and D2, respectively. ST-836 hydrochloride has the potential for Parkinson’s disease.
Specifications
Chemical Name or Material ST-836 hydrochloride
CAS 1415564-68-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H35ClN4OS
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (221.70 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (110.85 mM; Need ultrasonic)
SMILES [H]Cl.CCCN(CCN1CCN(C2=CC=CC=C2OC)CC1)C3CCC4=C(SC=N4)C3
Molecular Weight (g/mol) 451.07
Formula Weight 451.07
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration