Learn More
ST-193 hydrochloride, MedChemExpress
Product Code. 16494606
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494606 100 mg 100mg
16470759 1 mg 1mg
16463368 10 mg 10mg
16459896 50 mg 50mg
16432736 25 mg 25mg
16406097 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16494606 Supplier Medchem Express Supplier No. HY101441A
MedChemExpress ST-193 hydrochloride is a potent broad-spectrum arenavirus inhibitor; inhibits Guanarito, Junin, Lassa and Machupo virus with IC50 values of 0.44, 0.62, 1.4 and 3.1 nM, respectively.
ST-193 hydrochloride is a potent broad-spectrum arenavirus inhibitor
Specifications
Chemical Name or Material ST-193 hydrochloride
CAS 2320274-72-2
Color White
Purity Grade Notes Research
Molecular Formula C24H26ClN3O
Quantity 100 mg
Solubility Information DMSO : ≥ 150 mg/mL (367.70 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES CC(C1=CC=C(CNC2=CC=C3C(N=CN3C4=CC=C(OC)C=C4)=C2)C=C1)C.[H]Cl
Molecular Weight (g/mol) 407.94
Formula Weight 407.94
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus