missing translation for 'onlineSavingsMsg'
Learn More

SSR4 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16858883 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16858883 20 μL 20µL
16848883 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16858883 Supplier Proteintech Supplier No. 116552AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRAP proteins are part of a complex whose function is to bind calcium to the ER membrane and thereby regulate the retention of ER resident proteins. SSR4. also called TRAPD. is assumed to be involved in protein secretion. It is located in the Xq28 region.

Specifications

Antigen SSR4
Applications Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SSR4
Gene Accession No. P51571
Gene Alias SSR delta, SSR4, TRAP delta, TRAPD
Gene Symbols SSR4
Host Species Rabbit
Immunogen SSR4 Fusion Protein Ag2250
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6748
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.