missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSEA-5 Mouse anti-Human, BUV563, Clone: 8e11/SSEA-5 (also known as 8e11), BD Biosciences™
Description
SSEA-5 Monoclonal antibody specifically detects SSEA-5 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | SSEA-5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 8e11/SSEA-5 (also known as 8e11) |
| Conjugate | Brilliant Ultraviolet 563 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Stage-Specific Embryonic Antigen-5 |
| Host Species | Mouse |
| Immunogen | Human Undifferentiated Embryonic Stem Cells |
| Quantity | 50 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?