missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SSBP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16697614
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16697614 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16697614 Supplier Invitrogen™ Supplier No. PA5101140

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SSBP2.

SSBP2 is a member of a closely related, evolutionarily conserved, and ubiquitously expressed gene family, potential tumor suppressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SSBP2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SSBP2
Gene Accession No. P81877, Q9CYZ8
Gene Alias 1500004K09Rik; 2310079I02Rik; 9330163K02Rik; A830008M03Rik; AU067692; AW558684; DKFZp686F03273; HSPC116; RGD1562968; Sequence-specific single-stranded-DNA-binding protein 2; si:ch211-219m24.4; single stranded DNA binding protein 2; single-stranded DNA binding protein 2; single-stranded DNA-binding protein 2; SOSS-B2; Ssbp2; Ssdp2
Gene Symbols SSBP2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SSBP2(Accession P81877), corresponding to amino acid residues M1-S44.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23635, 66970
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.