Learn More
Invitrogen™ SRSF6 Recombinant Rabbit Monoclonal Antibody (24GB1825)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557266
This item is not returnable.
View return policy
Description
SRSF6 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SFRS6 is involved in mRNA splicing and may play a role in the determination of alternative splicing. It belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12. The protein encoded by this gene is involved in mRNA splicing and may play a role in the determination of alternative splicing. The encoded nuclear protein belongs to the splicing factor SR family and has been shown to bind with and modulate another member of the family, SFRS12.
Specifications
| SRSF6 | |
| Recombinant Monoclonal | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| G3V6S8, Q13247, Q3TWW8 | |
| Srsf6 | |
| A synthesized peptide derived from human SRSF6 (aa 164-199). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB1825 | |
| Unconjugated | |
| Srsf6 | |
| 1210001E11Rik; AI314910; AW146126; B52; epididymis secretory protein Li 91; HEL-S-91; LOW QUALITY PROTEIN: serine/arginine-rich splicing factor 6; MGC5045; pre-mRNA-splicing factor SRP55; serine and arginine rich splicing factor 6; serine/arginine-rich splicing factor 6; serine/arginine-rich splicing factor 6-like protein; Sfrs6; splicing factor SRp55; splicing factor SRp55 homolog; splicing factor SRp55-1; splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 6; SRP55; SRP55-1; SRSF6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 362264, 6431, 67996 | |
| -20°C | |
| Liquid |