missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SR140 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17128463
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17128463 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17128463 Supplier Invitrogen™ Supplier No. PA5110536

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GAPLEDVDGI PIDATPIDDL DGVPIKSLDD DLDGVPLDAT EDSKKNEPIF KVAPSKWEAV DESELEAQAV TTSKWELFDQ HEESEEEENQ NQEEESEDEE DTQSSKSEEH HLYSNPIKEE MTESKFSKYS EMSEEKRAKL REIELKVMKF QDELESGKRP KKPGQSFQEQ VEHYRDKLLQ REKEKELERE RERDKKDKEK LESRSKDKKE KDECTPTRKE RKRRHSTSPS PSRSSSGRRV KSPSPKSERS ERSERSHKES SRSRSSHKDS PRDVSKKAKR SPSGSRTPKR SRRSRSRSPK KSGKKSRSQS RSPHRSHKKS KKNKH.

U2SURP (U2 SnRNP Associated SURP Domain Containing) is a Protein Coding gene. Diseases associated with U2SURP include Retinitis Pigmentosa 67. Among its related pathways are Gene Expression and mRNA Splicing - Major Pathway. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and RNA binding. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SR140
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene U2SURP
Gene Accession No. O15042, Q6NV83
Gene Alias 140 kDa Ser/Arg-rich domain protein; 2610101N10Rik; AU023006; fSAPa; functional spliceosome-associated protein a; KIAA0332; mp:zf637-1-000918; RGD1307882; Ser/Arg-rich domain protein, 140 kDa; si:dkey-181m9.9; SR140; U2 associated SR140 protein; U2 snRNP associated SURP domain containing; U2 snRNP-associated SURP domain containing; U2 snRNP-associated SURP motif-containing protein; U2-associated protein SR140; u2-associated protein SR140-like protein; U2-associated SR140 protein; U2SURP; wu:fa03e02; wu:fb38b01; wu:fc75d03
Gene Symbols U2SURP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 705-1029 of human U2SURP (NP_001073884.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23350, 315903, 67958
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.