missing translation for 'onlineSavingsMsg'
Learn More

SR 9009, TRC

Product Code. 17987354
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17987354 10 mg 10mg
17959174 25 mg 25mg
17930005 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17987354 Supplier Toronto Research Chemicals Supplier No. TRCS68425510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1379686-30-2
Molecular Formula C20H24ClN3O4S
Molecular Weight (g/mol) 437.9
Synonym REV-ERB Agonist II
IUPAC Name 3-[[[(4-chlorophenyl)methyl][(5-nitro-2-thienyl)methyl]amino]methyl]-1-pyrrolidinecarboxylic acid ethyl ester
SMILES ClC(C=C1)=CC=C1CN(CC2CCN(C(OCC)=O)C2)CC3=CC=C([N+]([O-])=O)S3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H24ClN3O4S/c1-2-28-20(25)23-10-9-16(13-23)12-22(11-15-3-5-17(21)6-4-15)14-18-7-8-19(29-18)24(26)27/h3-8,16H,2,9-14H2,1H3
Formula Weight 437.1176
Percent Purity >95%
Chemical Name or Material SR9009
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.