missing translation for 'onlineSavingsMsg'
Learn More

Squalamine, MedChemExpress

Product Code. 16457058
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16457058

Brand: Medchem Express HY16468

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Squalamine(MSI-1256) is an aminosterol compound with potent broad spectrum antiviral activity.

Squalamine(MSI-1256) is an aminosterol compound with potent broad spectrum antiviral activity.

Specifications

Chemical Name or Material Squalamine
Physical Form Solid
CAS 148717-90-2
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C34H65N3O5S
For Use With (Application) COVID-19-anti-virus
Synonym MSI-1256
Solubility Information DMSO : 100 mg/mL (159.25 mM; Need ultrasonic)
SMILES C[C@@]12[C@](C[C@@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@@H](CC[C@H](C(C)C)OS(=O)(O)=O)C)([H])C[C@@H](NCCCNCCCCN)CC1
Molecular Weight (g/mol) 627.96
Formula Weight 627.96
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.