missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spinorphin TFA Salt, TRC
Specifications
Specifications
| Molecular Weight (g/mol) | 991.061 |
| Quantity | 2.5 mg |
| Shipping Condition | Room Temperature |
| Synonym | 6a-L-Threonine-myelopeptide 2∣Spinorphin (Cattle) (9CI) TFA Salt∣L-Leucyl-L-valyl-L-valyl-L-tyrosyl-L-prolyl-L-tryptophyl-L-threonine TFA Salt∣149:PN: US20120207734 SEQID: 148 Unclaimed Sequence TFA Salt∣1: PN: US20060177886 SEQID: 1 Unclaimed Sequence∣67: PN: WO2011020052 SEQID: 148 Claimed Protein TFA Salt∣7: PN: US6784155 SEQID: 7 Claimed Protein TFA Salt∣95: PN: WO2011146704 SEQID: 92 Unclaimed Protein TFA Salt∣LVV-Hemorphin-4 TFA Salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?