Learn More
Corning™ Spin-X™ Centrifuge Tube Insert Without Membrane
Product Code. 10486393 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
2 mL
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10486393 2 mL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10486393 Supplier Corning™ Supplier No. 9301
Only null left
Add to basket
Add to basket
Nonsterile and certified nonpyrogenic
  • Spin-X centrifuge tube filters consist of a membrane-containing filter unit within a microcentrifuge tube.
  • Uses: Removing bacteria, cells, and particles from liquids; HPLC sample preparation; DNA removal from agarose or acrylamide gels. Maximum RCF is 16,000 x g.
Order Info
Insert only (microfuge tube not included).
TRUSTED_SUSTAINABILITY
Specifications
Sterility Non-sterile
Volume (Metric) 2 mL