Learn More
BD Accuri™ Spherotech 8-Peak Validation Beads (FL1 - FL3)
Product Code. 12867880
Click to view available options
Quantity:
4 mL
Unit Size:
Each
Product Code. 12867880
Brand: BD 653144
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
8-Peak validation bead tests are designed to fluoresce in all channels on most flow cytometers and cell sorters, to measure subtle differences in fluorescence levels.
  • Check fluorescence sensitivity and resolution.
TRUSTED_SUSTAINABILITY
Specifications
Description 8-Peak validation beads
Product Type Validation Beads
For Use With (Equipment) Flow cytometers and Cell sorters
Quantity 4 mL
Size Distribution 4 mL