Learn More
Cole-Parmer™ XRF Prep-Aid™ Grinding Aids and Sample Binders
Product Code. 15360552 Shop All SPEX Certiprep Products
Click to view available options
Type:
Cellulose Binder
Paraffin Binder
Ultrabind
Quantity:
150 g
450 g
Unit Size:
150g
450g
Product Code. 15360552
Brand: Cole-Parmer™ 3644150
This item requires a price update prior to being ordered.
Request a Price for this item here.
Prepare samples for reproducible and contamination-free results with XRF Prep-Aid™ Grinding Aids and Sample Binders.
Ideal for samples which crumble or ablate after pressing.
  • Select binders from powder, tablet, or liquid form depending on your application.
  • High-quality sample preparation is the key to achieving consistent and reliable analytical results.
  • Powders are ideal for samples that crumble or ablate after pressing.
  • They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks.
  • Tablets are an efficient way to add binder to XRF sample disks.
  • Liquid easily mixes with the sample.
Specifications
For Use With (Equipment) SamplePrep Mixers, SamplePrep Mills
Type Ultrabind
For Use With (Application) Samples Which Crumble or Ablate After Pressing
Particle Size ≤20 μm
Quantity 150 g
Product Content Correction
Product Title
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt