Description
- Replaces polycarbonate center cylinders when sample contact with plastic is not acceptable.
- Made of nonmagnetic 440C stainless steel.
- Withstands liquid nitrogen temperatures.
- Autoclavable and reusable.
Specifications
Specifications
| Product Type | Center Cylinder |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| For Use With (Application) | Replaces Polycarbonate Cylinder When Sample Contact with Plastic Cannot be Used |
| For Use With (Equipment) | 6801 Large Grinding Vial Set (61043-10) |
| Material | Stainless Steel (440 C) |
| Packaging Quantity | 1 Ea. |